Find methods for your needs
HPLC analysis with charged aerosol detection of lauryl ether sulfates in shampoo
Description
This application shows the use of HPLC-CAD for analysis of lauryl ether sulfates in shampoo. Laureth sulfate (alkyl polyethoxy sulfate or lauryl ether sulfate (SLES)), not to be confused with lauryl sulfate (sodium dodecyl sulfate (SDS)), is a popular class of surfactants featuring good water solubility, high foam, and good cleaning properties. SLESs are used in many personal care products such as shampoo and hand soap. Different grades feature various mixtures of C12, C14 and C16 alkyl chains, with varying degrees of ethoxylation.| Market: | Chemical |
| Keywords: | Charged aerosol detector, Shampoo, Soap, Surfactants, CAD, foaming agent, hydrotrope |
| Matrix: | Shampoo |
| Author: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.