Find methods for your needs
HPAE-PAD Profiling of Maltodextrins using Dual Eluent Generation Cartridges
Description
The aim of this proof of concept study is to evaluate the analysis of variety of oligosaccharides using HPAE-PAD with dual eluent generation cartridges (EGC) on ICS-6000 HPIC system. The samples are maltodextrins in water, enzyme-hydrolyzed wheat arabinoxylan, and enzyme-hydrolyzed beechwood xylan. The report shows that the maltodextrin and the enzyme hydrolysates can be very well separated on a Dionex CarboPac PA200, 1X250 mm column set using a KOH/KMSA gradient delivered by dual EGC mode.| Market: | Food and Beverage |
| Keywords: | maltodextrin, oligosaccharides, HPAE-PAD, Dionex EGC 400 KOH, Dionex EGC 400 MSA, Dionex ICS-6000 HPIC system, Dual Eluent Generation, PA200-1mm column, AU150, TN 20, TN 70671, TN20, TN70671 |
| Matrix: | Water |
| Author: | Linda Lin; Michael Hvizd |
| Affiliation: | Thermo Fisher Scientific |
| Status: | Proof of Concept |
For Research Use Only. Not for use in diagnostic procedures.