Find methods for your needs
Separation of Eight Nucleotides and Six Organic Acids: KOH Gradient
Description
This report demonstrates the proof of concept to determine eight nucleotides and six organic acid metabolites by anion-exchange chromatography with suppressed conductivity detection. The metabolites were separated on the 2 mm i.d. Thermo Scientific Dionex IonPac AS11-HC-4um column, using electrolytically generated KOH gradient. These experiments were facilitated by the Thermo Scientific Dionex ICS-6000 HPIC system. Compare these results against those using methanol organic modifier (another record in AppsLab and in the attached report).| Market: | Metabolomics |
| Keywords: | AMP, ATP, Formate, Malate, Nucleotides, Sugar phosphates, dTTP, Citrate, Succinate, Isocitrate, Metabolites, RFIC, Oxalate, Reagent-Free IC, NADPH, metabolomics, ICS-6000, TCA cycle, IonPac AS11HC, phospho-sugars, dCMP, NAD, dGMP, UTP, ADRS 600, AN1137, AN622 |
| Matrix: | Standards in DI Water |
| Author: | Linda Lin and Michael Hzivd |
| Affiliation: | Thermo Fisher Scientific |
| Status: | Proof of Concept |
For Research Use Only. Not for use in diagnostic procedures.