Find methods for your needs
AN73105: Determination of Total Halogens, Sulfur, and Iodine in Liquefied Petroleum Gas by Pyrohydrolytic Combustion Ion Chromatography Be the first to rate this application
Description
Determinations of halogen and sulfur-containing compounds in liquified petroleum gas (LPG) is needed because these contaminants can foul the catalysts during processing. Pyrolytic combustion ion chromatography (CIC) is an ideal approach to eliminate the sample matrix and increase sample homogeneity. This application note demonstrates the determinations of total halo-benzene compounds and total dimethylsulfide in n-butane LPG as fluoride, chloride, bromide, iodide, and sulfate using pyrohydrolytic combustion coupled with IC.
Market: Petrochem, Oil and Gas; Chemical
Keywords: Eluent Generation, iodide, fluoride, RFIC, IC, IonPac AS20, Chloride, Sulfate, CIC, Sulfur, Chlorine, Fluorine, Bromine, Reagent-Free IC, Bromide, Integrion, LPG, combustion IC, TN72211, TN175, total halide, total sulfur, AS20, iodine, pyrohydrolytic combustion, TN 175, TN 72211, AN 73105, AN73105, TN000767, TN 000767
Matrix: LPG
Author: Terri Christison and Jeff Rohrer
Affiliation: Thermo Fisher Scientific, Sunnyvale, CA USA
Uploaded on 7/19/2019.
For Research Use Only. Not for use in diagnostic procedures.