Find methods for your needs
Novel Determination of Bromate in Aqueous Samples using 2-mm-Columns and UV-Detection, ISO 11206
Description
This proof of concept application determines trace bromate in aqueous samples by ISO 11206. The second scope of this work was to apply modern 2-mm-column technology to the derivatization approach described in the ISO method as well as in Thermo Fisher Scientific Technical Note 116. Both those documents use 4-mm-columns. Using 2 mm columns reduced eluent consumption and hence waste.| Market: | Environmental; Water Analysis |
| Keywords: | Bromate, UV, aqueous samples, ISO 11206, TN116, eWorkflow |
| Matrix: | Drinking water |
| Author: | Ralph Neufang, Julian Görl, Detlef Jensen |
| Affiliation: | Thermo Fisher Scientific |
| Status: | Proof of Concept |
For Research Use Only. Not for use in diagnostic procedures.