Find methods for your needs
CAN72995: A More Cost-Efficient Extraction Method for Polycyclic Aromatic Hydrocarbons (PAH) in Sediments and Soils using Accelerated Solvent Extraction (ASE)
Description
ASE is commonly used to extract polycyclic aromatic compounds (PAC) from solid matrices. The development of ASE for PAHs can be traced to the 1990s. Recently, a one-step ASE method that negated the need for adsorption chromatography was reported to extract PAHs from marine sediments. Inspired by the previous work, the Centre for Oil and Gas Research and Development (COGRAD) decided to further investigate the one-step ASE method for PAHs on two standard reference materials. The goal was to verify the suitability of the one-step method and its applicability to improve laboratory efficiency.| Market: | Environmental |
| Keywords: | ASE 350, US EPA, APAHs, CAN 72995, HPACs, PACs |
| Matrix: | Soil |
| Author: | Zhe Xia, Nicole Klaassen, Ifeoluwa Idowu, and Gregg Tomy |
| Affiliation: | University of Manitoba, Winnipeg, MB, Canada |
For Research Use Only. Not for use in diagnostic procedures.