Find methods for your needs
Column Performance Test Using the Quality Assurance Report (QAR) Conditions for the 2 mm Dionex IonPac CS20 column
Description
Before running any samples, Thermo Scientific recommends that you first confirm the performance of the column by reproducing the lot validation report chromatogram shipped with column. Compare your results with the one reported in the quality assurance report (QAR). This record provides an eWorkflow for executing the QAR method on an HPIC system. See column manual for detail. This column is suitable for Group I and Group II cations, alkylamines and alkanolamines.| Market: | Chemical; Petrochem, Oil and Gas |
| Keywords: | Amines, Magnesium, Lithium, RFIC, Potassium, IC, Calcium, Sodium, Ammonium, QAR, CS20, IonPac CS20, Dionex IonPac CS20 |
| Matrix: | Water |
| Author: | Terri Christison, John Guajardo, and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.