Find methods for your needs
Fast and easy separation of 23 drugs of abuse including high, stable resolution of isobaric opioids from human urine by UHPLC-MS/MS
Description
Fast and Easy separation of 23 drugs of abuse, including the adequate and stable resolution of isobaric opioids from human urine. The new Thermo Scientific™ Accucore™ Biphenyl solid core column is used in combination with the Thermo Scientific™ Horizon™ UHPLC system and Thermo Scientific™ TSQ Quantiva™ MS/MS system.| Market: | Pharma |
| Keywords: | benzodiazepines, drugs of abuse, Solid core, Urine, Opioids, TSQ Quantiva MS, Vanquish Horizon, Accucore column, cannibinoids, isobaric separation, biphenyl, UHPLC system |
| Matrix: | Human urine |
| Author: | Kean Woodmansey, Jon Bardsley, Stacy Tremintin |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.