Find methods for your needs
AU72996: Determination of Potential Sulfate in Denatured Ethanol using Modified ASTM D7328 Method
Description
This Application Updated validated the modified ASTM D7328 procedure for determining potential sulfate in denatured ethanol using a compact high-pressure ion chromatography (HPIC™) system with suppressed conductivity detection.| Market: | Energy |
| Keywords: | biofuel, Dionex Integrion HPIC, ASTM D7328, Thermo Scientific Dionex IonPac AS22 Column, AU194 |
| Matrix: | Biofuel, Denatured ethanol |
| Author: | Beibei Huang and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.