Find methods for your needs
Routine-grade quantitative performance of TriPlus 500 Headspace Autosampler coupled to TRACE 1310 GC-FID
Description
In this technical note, a Thermo Scientific™ TriPlus™ 500 Headspace Autosampler was coupled to a Thermo Scientific™ TRACE™ 1310 Gas Chromatograph equipped with a Thermo Scientific™ Instant Connect Split/Splitless (SSL) Injector and a Thermo Scientific™ Instant Connect FID to assess the linearity for Class 1, Class 2A and Class 2B residual solvents.| Market: | Pharma |
| Keywords: | FID, GC, Pharmaceuticals, Headspace, Residual solvents, Quantification, Gas Chromatography, Linearity, Flame ionization detector, USP <467>, HS, MDL, method detection limit, routine grade, TriPlus 500 headspace autosampler |
| Matrix: | Water |
| Author: | Giulia Riccardino and Cristian Cojocariu |
| Affiliation: | Thermo Fisher Scientific, Runcorn |
For Research Use Only. Not for use in diagnostic procedures.