Find methods for your needs
Detection and quantification of potential fragrance allergens in complex matrices using GC Orbitrap MS technology
Description
In this work the performance of an Orbitrap-based GC-MS was tested for the analysis of 57 potential fragrance allergens (60 analytes including isomers). Using the unparalleled high resolving power, linear dynamic range, and sensitivity, these potentially allergenic compounds were confidently detected, identified, and quantified at low to high concentration levels in a robust manner and from possibly complex samples.| Market: | Chemical |
| Keywords: | GC-MS, High resolution, Chromeleon CDS, Quantification, Linearity, Exact Mass, GC Orbitrap, allergens, Fragrance |
| Matrix: | fragrance, perfumes |
| Author: | Cristian Cojocariu1, Emilie Belhassen2, Richard Law1, and Philippe Merle2 |
| Affiliation: | 1Thermo Fisher Scientific, Runcorn, UK 2Firmenich SA, Geneva, Switzerland |
For Research Use Only. Not for use in diagnostic procedures.