Find methods for your needs
Completely cryogen-free monitoring of ozone precursors, air toxics and oxygenated volatile organic compounds in ambient air in a single run Be the first to rate this application
air pollutants ambient air
Description
Using an analytical system comprised of Markes CIA Advantage-xr canister autosampler, Kori-xr water removal device, and UNITY-xr thermal desorber coupled to a dual-column Thermo Scientific™ ISQ™ 7000 single quadrupole GC-MS, for the analysis of ozone precursors, air toxics and oxygenated volatile organic compounds in ambient air.
Market: Environmental
Keywords: FID, GC, Gas Chromatography, air toxics, CIA Advantage-xr canister autosampler, cryogen-free monitoring, Flame ionization detector, ISQ 7000, Kori-xr water removal device, OVOCs, Oxygenated volatile organic compounds, ozone precursors, PAMS, photo assessment monitoring scheme, signal quadrupole mass spectrometer, TO-15, UNITY-xr thermal desorber
Matrix: ambient air
Author: Jane Cooper(1) Laura Miles (2) Natasha D Spadafora (2)
Affiliation: (1) Thermo Fisher Scientific, Runcorn, UK (2) Markes International Ltd, Llantrisant, UK
Uploaded on 4/24/2019.
For Research Use Only. Not for use in diagnostic procedures.