Find methods for your needs
Completely cryogen-free monitoring of ozone precursors, air toxics and oxygenated volatile organic compounds in ambient air in a single run
Description
Using an analytical system comprised of Markes CIA Advantage-xr canister autosampler, Kori-xr water removal device, and UNITY-xr thermal desorber coupled to a dual-column Thermo Scientific™ ISQ™ 7000 single quadrupole GC-MS, for the analysis of ozone precursors, air toxics and oxygenated volatile organic compounds in ambient air.| Market: | Environmental |
| Keywords: | FID, GC, Gas Chromatography, air toxics, CIA Advantage-xr canister autosampler, cryogen-free monitoring, Flame ionization detector, ISQ 7000, Kori-xr water removal device, OVOCs, Oxygenated volatile organic compounds, ozone precursors, PAMS, photo assessment monitoring scheme, signal quadrupole mass spectrometer, TO-15, UNITY-xr thermal desorber |
| Matrix: | ambient air |
| Author: | Jane Cooper(1) Laura Miles (2) Natasha D Spadafora (2) |
| Affiliation: | (1) Thermo Fisher Scientific, Runcorn, UK (2) Markes International Ltd, Llantrisant, UK |
For Research Use Only. Not for use in diagnostic procedures.