Find methods for your needs
Rapid analysis of chlorhexidine digluconate using a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of chlorhexidine digluconate. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column, with UV detection at 260 nm.| Market: | Pharma |
| Keywords: | Chlorhexidine digluconate, HPLC, Hypersil GOLD, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.