Find methods for your needs
Transfer of an EP method for mebendazole from a Waters Acquity UPLC system to a Vanquish Horizon UHPLC system
Description
The current application demonstrates the straightforward transfer of an EP monograph HPLC method from a Waters Acquity UPLC system to a Thermo Scientific Vanquish Horizon UHPLC system. During the transfer flexible thermostatting options provided by the Vanquish platform help to mimic the actual conditions at the original system. Substantial time and solvent savings are obtainable by speeding up the conventional HPLC method to UHPLC conditions without sacrificing chromatographic performance.| Market: | Pharma |
| Keywords: | HPLC method transfer, European Pharmacopoeia, mebendazole, Vanquish Horizon UHPLC system, Waters Acquity UPLC system |
| Matrix: | N/A |
| Author: | Maria Grübner |
| Affiliation: | Thermo Fisher Scientific, Germering, Germany |