Find methods for your needs
The VetDrugs Explorer Collection: screening and quantitation of multi-class veterinary drug residues in animal matrices with a comprehensive workflow solution
# | Compound Name | Compound Class | RT (min) |
---|---|---|---|
1 | 2,4,6-Triamino-pyrimidine-5-carbonitrile | Dycyclanil Marker Residue | 0.75 |
2 | Cyromazine | Ectoparasiticide | 0.82 |
3 | Sulfaguanadine | Antibiotic (sulfonamide) | 1.11 |
4 | Sulfanilamide | Antibiotic (sulfonamide) | 1.12 |
5 | Desacetylcefapirin | Metabolite | 1.84 |
6 | Hydroxymetronidazole | Metronidazole metabolite | 2.3 |
7 | Dicyclanil | Insecticide/larvacide | 2.58 |
8 | Terbutaline | β-agonist | 3.42 |
9 | Zilpaterol | β-agonist | 3.67 |
10 | Metronidazole | Antibiotic (nitroimidazole) | 3.74 |
11 | Salbutamol | β-agonist | 3.79 |
12 | Furaltadone | Antibiotic (nitrofuran) | 3.9 |
13 | Ronidazole | Coccidiostat | 4.02 |
14 | Dimetridazole | Coccidiostat (antiprotozoal) | 4.07 |
15 | Cimbuterol | β-agonist | 4.17 |
16 | Sulfadiazine | Antibiotic (sulfonamide) | 4.17 |
17 | 5-hydroxythiabendazole | Thiabendazole metabolite | 4.21 |
18 | Tildipirosin | Antibiotic (macrolide) | 4.23 |
19 | Cefapirin | Antibiotic (b-lactam) | 4.25 |
20 | Clopidol | Coccidiostat (antiprotozoal | 4.34 |
21 | Sulfathiozole | Antibiotic (sulfonamide) | 4.44 |
22 | Levamisole | Anti parasitic worm | 4.45 |
23 | Lincomycin | Antibiotic (macrolide) | 4.45 |
24 | Cefquinome | Antibiotic (b-lactam) | 4.52 |
25 | Thiabendazole | Antiparasitic; antiroundworm | 4.52 |
26 | Marbofloxacin | Antibiotic (quinolone) | 4.54 |
27 | Sulfapyridine | Antibiotic (sulfonamide) | 4.57 |
28 | Trimethoprim | Antibiotic | 4.58 |
29 | Epitetracycline | Antibiotic (tetracycline) | 4.62 |
30 | Norfloxacin | Antibiotic (quinolone) | 4.67 |
31 | Epioxytetracycline | Antibiotic (tetracycline) | 4.68 |
32 | Albendazole 2-aminosulfone | Albendazole metab | 4.71 |
33 | Cephalonium | Antibiotic (b-lactam) | 4.71 |
34 | Sulfamerazine | Antibiotic (sulfonamide) | 4.72 |
35 | Ciprofloxacin | Antibiotic | 4.75 |
36 | Clenproperol | β-agonist | 4.8 |
37 | Oxytetracycline | Antibiotic (tetracycline) | 4.83 |
38 | Azaperol | Azaperone metabolite | 4.84 |
39 | Danofloxacin | Antibiotic | 4.84 |
40 | Ractopamine | β-agonist | 4.84 |
41 | Lomefloxacin | Antibiotic (quinolone) | 4.88 |
42 | Enrofloxacin | Antibiotic (quinolone) | 4.92 |
43 | Tetracycline | Antibiotic (tetracycline) | 4.94 |
44 | Thiamphenicol | Antibiotic | 5.02 |
45 | Sulfamethizole | Antibiotic (sulfonamide) | 5.13 |
46 | Xylazine | Sedative and anesthetic | 5.14 |
47 | Azaperone | Sedative and antiemetic | 5.17 |
48 | Sulfamethazine | Antibiotic (sulfonamide) 0.60 86.0 | 5.17 |
49 | Clenbuterol | β-agonist | 5.22 |
50 | Difloxacin | Antibiotic (quinolone) | 5.22 |
51 | Salmeterol | β-agonist | 5.22 |
52 | Sarafloxacin | Antibiotic (quinolone) | 5.22 |
53 | Sulfamethoxypyridazine | Antibiotic (sulfonamide) | 5.23 |
54 | Neospiramycin I | Major metabolite of Spiramycin I | 5.24 |
55 | Amoxicillin | Antibiotic (b-lactam) | 5.25 |
56 | Ampicillin | Antibiotic (b-lactam) | 5.25 |
57 | Cefazolin | Antibiotic (b-lactam) | 5.29 |
58 | Dapsone | Antibiotic | 5.3 |
59 | Epichlortetracycline | Antibiotic (tetracycline) | 5.33 |
60 | Hydroxyipronidazole | Ipronidazole metabolite | 5.36 |
61 | Albendazole sulfoxide | Albendazole metab | 5.55 |
62 | Mebendazole amine | Antihelmintic | 5.57 |
63 | Spiramycin I | Antibiotic (macrolide) | 5.57 |
64 | Sulfachlorpyridazine | Antibiotic (sulfonamide) | 5.6 |
65 | Sulfamonomethoxine | Antibiotic (sulfonamide) | 5.62 |
66 | Isoxsuprine | β-agonist | 5.66 |
67 | Azure B | Aquaculture (Dye) | 5.71 |
68 | Clenpenterol | β-agonist | 5.71 |
69 | Mebendazol-hydroxy | Mebendazole metabolite | 5.72 |
70 | Pararosaniline base | Aquaculture (Dye) | 5.72 |
71 | Chlorpromazine | Sedative and antiemetic | 5.73 |
72 | Mabuterol | β-agonist | 5.74 |
73 | Flubendazole amine | Flubendazole metabolite | 5.78 |
74 | Cefoperazone | Antibiotic (b-lactam) | 5.79 |
75 | Carazolol | Beta blocker | 5.8 |
76 | Sulfamethoxazole | Antibiotic (sulfonamide) | 5.8 |
77 | Sulfadoxine | Antibiotic (sulfonamide) | 5.85 |
78 | Methylene Blue | Aquaculture (Dye) | 5.89 |
79 | Gamithromycin | Antibiotic (macrolide) | 5.94 |
80 | Pirlimycin | Antibiotic (lincosamide) | 5.96 |
81 | Sulfisoxazole | Antibiotic (sulfonamide) | 6.01 |
82 | Doxycycline | Antibiotic (tetracycline) | 6.07 |
83 | Halofuginone | Coccidiostat (antiprotozoal) | 6.07 |
84 | Oxibendazole | Antihelmintic | 6.15 |
85 | Ipronidazole | Coccidiostat (antiprotozoal) | 6.17 |
86 | Mapenterol | β-agonist | 6.19 |
87 | Albendazole sulfone | Albendazole metab | 6.2 |
88 | Tilmicosin | Antibiotic (macrolide) | 6.2 |
89 | Clorsulon | Antihelmintic | 6.28 |
90 | Oxfendazole | Antihelmintic | 6.37 |
91 | Oxolinic acid | Antibiotic (quinolone) | 6.37 |
92 | Derquantel | Antihelmintic | 6.53 |
93 | Ceftiofur | Antibiotic (b-lactam) | 6.56 |
94 | Sulfadimethoxine | Antibiotic (sulfonamide) | 6.56 |
95 | Sulfaquinoxaline | Antibiotic (sulfonamide) | 6.63 |
96 | Haloperidol | Sedative and antiemetic | 6.8 |
97 | Acepromazine | Sedative and antiemetic | 6.91 |
98 | Fenbendazole Sulfone | Fenbendazole metabolite | 7.02 |
99 | Erythromycin | Antibiotic (macrolide) | 7.04 |
100 | Leucocrystal violet | Aquaculture (Dye) | 7.06 |
101 | New methylene blue | Aquaculture (Dye) | 7.08 |
102 | Tylosin A | Antibiotic | 7.24 |
103 | Nalidixic acid | Antibiotic (quinolone) | 7.32 |
104 | Mebendazole | β-agonist | 7.34 |
105 | Albendazole | Anti-parasitic worm | 7.41 |
106 | Penicillin G | Antibiotic b-lactam | 7.43 |
107 | Propionylpromazine | Sedative and antiemetic | 7.43 |
108 | Tylosin-3-acetate | Metabolite | 7.46 |
109 | Nitroxynil | Antihelmintic/antiparasitic | 7.49 |
110 | Flumequine | Antibiotic (quinolone) | 7.52 |
111 | Chlortetracycline | Antibiotic (tetracycline) | 7.6 |
112 | Flubendazole | Antihelmintic | 7.64 |
113 | Nile blue A | Aquaculture (Dye) | 7.66 |
114 | Penicillin V | Antibiotic b-lactam | 7.78 |
115 | Malachite green | Aquaculture (Dye) | 7.79 |
116 | Valnemulin | Antibiotic (pleuromutilin) | 7.92 |
117 | Betamethazone | Steroid | 7.95 |
118 | Dexamethasone | Steroid anti-inflammatory | 7.95 |
119 | Josamycin | Antibiotic (macrolide) | 7.99 |
120 | Cloxacillin | Antibiotic (b-lactam) | 8.21 |
121 | Oxacillin | Antihelmintic | 8.21 |
122 | Firocoxib | NSAID | 8.34 |
123 | Fenbendazole | Antihelmintic | 8.4 |
124 | Virginiamycin_M1 | Antibiotic (streptogramin) | 8.41 |
125 | Rhodamine-6g | Aquaculture | 8.44 |
126 | Dicloxacillin | Antibiotic (b-lactam) | 8.46 |
127 | Meloxicam | NSAID | 8.51 |
128 | Crystal violet | Aquaculture (Dye) | 8.54 |
129 | Ketoprofen | NSAID | 8.56 |
130 | Tylvalosin | Antibiotic (macrolide) | 8.56 |
131 | 5-Hydroxyflunixin | Flunixin metabolite | 8.65 |
132 | Rifampicin | Antibiotic | 8.79 |
133 | Testosterone | Steroid hormones | 8.91 |
134 | Leucomalachite green | Aquaculture (Dye) | 8.93 |
135 | Brilliant Green | Fungicide | 9.01 |
136 | Flunixin | NSAID | 9.07 |
137 | 1,3-bis(4-nitrophenyl) urea | Coccidiostat | 9.13 |
138 | Victoria blue b | Aquaculture (Dye) | 9.21 |
139 | Rifaximin | Antibiotic | 9.49 |
140 | Carprofen | NSAID | 9.51 |
141 | Altrenogest | Steroid | 9.52 |
142 | Diflubenzuron | Insecticide | 9.63 |
143 | Phenylbutazone | NSAID | 9.64 |
144 | Febantel | Antihelmintic | 9.65 |
145 | Diclofenac | NSAID | 9.72 |
146 | Triclabendazole sulfoxide | Triclabendazole metabolite | 9.73 |
147 | Niflumic acid | NSAID | 9.77 |
148 | Diclazuril | Coccidiostat (antiprotozoal) | 9.78 |
149 | Emamectin B1a | Antihelmintic | 9.79 |
150 | Victoria blue bo | Aquaculture (Dye) | 9.79 |
151 | Triclabendazole sulfone | Triclabendazole metabolite | 9.88 |
152 | Ethyl violet | Aquaculture (Dye) | 9.98 |
153 | Megestrol acetate | Steroidal progestin | 10.02 |
154 | Chlormadinone acetate | Progestin | 10.04 |
155 | Medroxyprogesterone acetate | β-agonist | 10.12 |
156 | Oxyclozanide | Antihelmintic | 10.14 |
157 | Triclabendazole | Antihelmintic | 10.19 |
158 | Tolfenamic acid | NSAID | 10.58 |
159 | Teflubenzuron | Aquaculture | 10.75 |
160 | Fluazuron | Insecticide | 11.05 |
161 | Decoquinate | Coccidiostat (antiprotozoal) | 11.38 |
162 | Eprinomectin B1a | Antihelmintic | 11.59 |
163 | Abamectin B1a | Antihelmintic | 11.76 |
164 | Closantel | Antihelmintic | 11.89 |
165 | Doramectin | Antihelmintic | 12 |
166 | Lasalocid | Coccidiostat (antiprotozoal) | 12.12 |
167 | Moxidectin | Antihelmintic | 12.13 |
168 | Monensin | Antibiotic | 12.22 |
169 | Rafoxanide | Antihelmintic | 12.24 |
170 | Ivermectin B1a | Antihelmintic | 12.34 |
171 | Salinomycin | Coccidiostat (ionophore) | 12.45 |
172 | Maduramicin | Coccidiostat (antiprotozoal) | 12.53 |
173 | Narasin | Coccidiostat (antiprotozoal) | 12.71 |
Description
A robust, sensitive, and reliable multi-residue LC-MS/MS method workflow solution that uses the Thermo Scientific TSQ Altis triple quadrupole mass spectrometer provides analysis and quantitation of more than 160 veterinary drugs with the Thermo Scientific VetDrugs Explorer Collection solution in salmon (fillet), bovine muscle, and milk. The complete instrument and data processing method including SRM settings is included with the VetDrugs Explorer Collection start-to-finish workflow solution. In 17 minutes, the target veterinary drugs were detected and quantified in food matrices.Market: | Food and Beverage |
Keywords: | TraceFinder Software, triple quadrupole mass spectrometry, TSQ Altis MS, VetDrugs Explorer Collection, Veterinary Medicines |
Matrix: | Bovine muscle, Milk, Salmon |
Author: | Ed George, Jonathan Beck, Charles Yang, Dave Borts, Viet Dang; George Stubbings, Danny Chan |
Affiliation: | Thermo Fisher Scientific, San Jose, CA; Veterinary Diagnostic Laboratory, College of Veterinary Medicine, Iowa State University, Ames, IA; Fera Science Ltd., York, UK |
For Research Use Only. Not for use in diagnostic procedures.