Find methods for your needs
AN72911: Fast Determination of Oxyhalides and Bromide in Drinking Water using a Compact Ion Chromatography System Coupled with a Single Quadrupole Mass Spectrometer
Description
This study demonstrates that oxyhalides and bromide can be determined accurately in municipal drinking water and bottled water using a fast isocratic 15-min IC-MS method. The method provides an advantage over the traditional IC-CD method concerning sensitivity and selectivity, offering positive analyte-identification, particularly for complex samples.| Market: | Environmental |
| Keywords: | Bromate, Chlorate, Chlorite, Disinfection Byproducts, isocratic, IC-MS, Drinking water, Dionex IonPac AS19-4μm column, Dionex Integrion HPIC, US EPA Method 300.1 Part B, AERS 500e suppressor, ISQ-EC |
| Matrix: | Water |
| Author: | Jingli Hu; Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.