Find methods for your needs
Profiling of Sialylated Oligosaccharides in Mammalian Milk Using Online Solid Phase Extraction-Hydrophilic Interaction Chromatography Coupled with Negative-Ion Electrospray MS
Description
Sialylated oligosaccharides are important components in mammalian milk. They play a key role in the health and growth of infants by helping to shape up infant’s gastrointestinal microbiota and defense against infection by various pathogenic agents. A detailed knowledge of the structures, compositions, and quantities of the sialylated milk oligosaccharides (SMOs) is a prerequisite for understanding their biological roles. However, because of the presence of very large amounts of lactose and neutral oligosaccharides, accurate analysis of SMOs is difficult. An online SPE-HILIC-MS was developped.| Market: | Food and Beverage |
| Matrix: | Acetonitrile, Water |
For Research Use Only. Not for use in diagnostic procedures.