Find methods for your needs
Dionex CarboPac PA1 2 mm Column Performance Test Using its QAR Method
Description
Before running any samples, Thermo Scientific recommends that you first confirm the performance of the column by reproducing the lot validation report chromatogram shipped with column. Compare your results with the one reported in the quality assurance report. At least three injections should be made. This record provides an eWorkflow for executing the QAR method on an ICS-5000+ system. An ICS-6000 can be used for this application.Market: | Food and Beverage; BioPharma |
Keywords: | CarboPac PA1, Electrochemical detection, food, Glucose, glycoprotein, Monosaccharides, Maltose, Sucrose, Sugars, fructose, HPAE-PAD, CarboPac PA1 column manual, TN 20, TN 70671, TN20, TN70671 |
Matrix: | DI water |
Author: | Manali Aggrawal; Jeff Rohrer |
Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |