Find methods for your needs
Fast separation of Organochlorine Pesticides (OCPs) using GC 0.32 mmID dual-column
Description
Thermo Scientific TraceGOLD TG-OCP columns offer a fully separation of organochlorine pesticides in less than 7 min. The TG-OCP I and TG-OCP II column pair is designed with different selectivity, which changes several elution order to fill the conformational requirement of EPA methods.Market: | Environmental; Food and Beverage |
Keywords: | Organochlorine Pesticides (OCPs) |
For Research Use Only. Not for use in diagnostic procedures.