Find methods for your needs
Separation of benzodiazepines using highly inert 5% diphenyl/95% dimethyl polysiloxane column
Description
Thermo Scientific TraceGOLD TG-5SilMS columns offer improved inertness compared to typical 5% diphenyl/95% dimethyl polysiloxane columns. Excellent peak shapes of benzodiazepines and complete separation were obtained less than 8 min| Market: | Toxicology; Forensics |