Find methods for your needs
Assay analysis of keto-analogues of essential amino acid calcium salt
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the assay analysis of keto-analogues of amino acid calcium salt. The analysis was performed on a Thermo Scientific Acclaim 120 C18 HPLC column. The separation results are optimum for assay analysis of keto-analalogues of amino acid calcium salt.| Market: | Pharma |
| Matrix: | N.A. |
| Author: | krishna rao dara |
| Affiliation: | thermo Scientific, CoE Lab, Ahmedabad, India |