Find methods for your needs
A comparison study of total acidity methods for the analysis of wine
Description
Colorimetric determination of Total Acids provides a rapid, user-friendly way of analyzing total acidity from alcoholic beverages such as white wine, red wine, cider, and liqueur. Along with the Total Acids method, other colorimetric or enzymatic methods, for example, glucose, fructose, acetaldehyde, total polyphenol, calcium, sulfur dioxide, can be run simultaneously on the analyzer.| Market: | Food and Beverage |
| Keywords: | Acids, liqueur, wine, discrete analysis, cider, acidity, Automated photometric analysis, titration |
| Matrix: | Wine |
| Author: | Sari Hartikainen; Mari Kiviluoma; Leena Kaski; Annu Suoniemi-Kahara |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.