Find methods for your needs
Correlation of an automated discrete analysis sulfur dioxide method to standardized para-rosaniline methods in the analysis of beer
Description
The SO2 Total method showed results similar to the EBC (9.25.3) p-rosaniline method across a variety of samples. The Gallery method was easy to use and provided a robust method for SO2 Total measurement for both cider and beer samples. Ready-to-use Thermo Scientific system reagents eliminated reagent preparation, which saved time, and the volume optimized methods minimized reagent waste. Thermo Scientific system reagents along with the fully automated Thermo Scientific discrete analyzers provide a total system solution for food and beverage analysis.| Market: | Food and Beverage |
| Keywords: | Brewing, Flavor, Beer, ASBC, cider, DTNB, EBC, para-rosaniline, Photometric, quality, stale beer, sulfur dioxide |
| Matrix: | Beer, Cider |
| Author: | Mari Klemm; Paula Ranta; Liisa Otama; Annu Suoniemi-Kahara |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.