Find methods for your needs
AN72808: Determination of Organic Acids in Herbal Beverages using a Compact Ion Chromatography System Coupled with Mass Spectrometry
Description
This work uses IC with suppressed conductivity and mass spectrometry detection for organic acid determinations. This dual detection approach increases the information available from the sample. Samples were separated with a high-resolution Thermo Scientific™ Dionex™ IonPac™ AS11-HC-4μm column set using a Thermo Scientific™ Dionex™ Integrion™ HPIC™ system with suppressed conductivity detection, and an ISQ EC MS. Co-eluting organic acids can be accurately quantified with mass spectrometric detection, eliminating the need for the addition of methanol to the hydroxide eluent to enhance separation.| Market: | Food and Beverage |
| Keywords: | Organic acid, RFIC System, ISQ EC Single Quadrupole Mass Spectrometer, Dionex Integrion, Dionex IonPac AS11-HC-4μm column, ion suppression |
| Matrix: | Beverage |
| Author: | Beibei Huang; Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.