Find methods for your needs
AN72765: Determination of Anionic Polar Pesticides and Oxyhalides in Beer and Strawberry Samples using IC-HRAM-MS
Description
Food safety and perceived health risks from residual agricultural chemicals are ongoing public concerns, and these chemicals are under increasing regulatory scrutiny. The polar pesticides are one category, which include ionic post-emergent and desiccant herbicides, fungicides, growth-regulating chemicals, and the resultant metabolites of those compounds (Table 1 in the attached application note).| Market: | Food and Beverage |
| Keywords: | Herbicide, RFIC, ion chromatography, IC, DBP, mass spectrometry, IC-MS, IonPac AS19, Suppressed Conductivity, Q Exactive, Integrion, Fungicides, polar pesticides, QuPPe, IC-HRAM MS, disinfectant byproducts, oxyhalide |
| Matrix: | diluted beer, strawberry extracts |
| Author: | Terri Christison; Luca Gerardo, Jonathan Beck, and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.