Find methods for your needs
Improved analysis of polycyclic aromatic hydrocarbons (PAHs) using a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of polycyclic aromatic hydrocarbons (PAHs). The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column, with UV detection at 269 nm.| Market: | Chemical |
| Keywords: | Anthracene, Chrysene, Flourene, HPLC, Hypersil GOLD, Naphthalene, PAHs, Phenanthrene, Polycyclic Aromatic Hydrocarbon (PAH), Pyrene, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.