Find methods for your needs
Method transfer of a USP derived acetaminophen assay from an Agilent 1260 Infinity system to an UltiMate 3000 SD system and a Vanquish Flex UHPLC system
Description
This application demonstrates the straightforward transfer of an analytical HPLC method from an Agilent 1260 Infinity instrument to a Thermo Scientific UltiMate 3000 HPLC system and a Thermo Scientific Vanquish Flex UHPLC system with the example of a USP derived assay for the analysis of acetaminophen and its impurities.Market: | Pharma |
Keywords: | Acetaminophen, HPLC method transfer, Agilent 1260 Infinity quaternary system, UltiMate 3000 Standard quaternary system, Vanquish Flex quaternary system, gradient delay volume |
Matrix: | Methanol |
Author: | Maria Grübner, Carsten Paul, Frank Steiner |
Affiliation: | Thermo Fisher Scientific, Germering, Germany |