Find methods for your needs
Method transfer of a USP-derived acetaminophen assay from an UltiMate 3000 SD system to a Vanquish Flex UHPLC system
Description
This application demonstrates the straightforward transfer of an analytical HPLC method from a Thermo Scientific UltiMate 3000 HPLC system to a Thermo Scientific Vanquish Flex UHPLC system demonstrated with a USP derived assay for the analysis of acetaminophen and its impurities.| Market: | Pharma |
| Keywords: | Acetaminophen, gradient delay volume, HPLC method transfer, UltiMate 3000 Standard quaternary system, Vanquish Flex quaternary system |
| Matrix: | Methanol |
| Author: | Maria Grübner, Carsten Paul, Frank Steiner |
| Affiliation: | Thermo Fisher Scientific, Germering, Germany |