Find methods for your needs
TN72778: Determination of Inorganic Anions in Drinking Water
Description
The determination of inorganic anions in municipal drinking, waste, and bottled waters according to the U.S. Environmental Protection Agency (EPA) Methods 300.0 and 300.1 is one of the most popular and widely used ion chromatography (IC) methods. This technical note provides detailed instructions for system set up and operation for determining inorganic anions in samples such as drinking water. We used Thermo Scientific™ Dionex™ Aquion™ ion chromatography system with a Thermo Scientific™ Dionex™ AS-DV Autosampler to determine seven anions in drinking water samples.| Market: | Environmental |
| Keywords: | fluoride, inorganic anions, Nitrate, Nitrite, Chloride, phosphate, Sulfate, carbonate/bicarbonate eluent, Dionex IonPac AS22 column, Drinking water, Bromide, AS-DV autosampler, Dionex Aquion IC system, U.S. EPA Method 300.1, bottle water |
| Matrix: | Water |
| Author: | Manali Aggrawal, Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |