Find methods for your needs
Simple charge variant profile comparison of an innovator monoclonal antibody and a biosimilar candidate
Description
To demonstrate the effectiveness of a simple pH gradient/ion-exchange chromatography workflow approach to the characterization of charge variant profiles of an innovator molecule (cetuximab) and a candidate biosimilar. To show the assay is simple, reproducible, easily optimized and resolves variants effectively.| Market: | BioPharma |
| Keywords: | Monoclonal antibody, protein characterization, Biopharmaceutical, IgG, mAb, cation exchange chromatography, Cetuximab, Innovator, Biosimilar, pH gradient buffer, charge variant analysis, NIBRT, Critical quality attribute, QA/QC, Biotherapeutic, CX-1 pH buffer, CEX, intact protein analysis, ion-exchange chromatography |
| Matrix: | Water |
| Author: | NIBRT |
For Research Use Only. Not for use in diagnostic procedures.