Find methods for your needs
1 mm Dionex CarboPac PA200 column Performance Test Using its QAR Method with Dual Eluent Generator Cartridge Mode
Description
Before running any samples, Thermo Scientific recommends that you first confirm the performance of the column by reproducing the lot validation report chromatogram shipped with column. Compare your results with the one reported in the quality assurance report. At least three injections should be made. This record provides an eWorkflow for executing the QAR method on an ICS-6000 system using Dual EGC mode.| Market: | Food and Beverage; BioPharma |
| Keywords: | Electrochemical detection, PAD, HPAE-PAD, Quality Assurance Report (QAR), Dionex ICS-6000, complex carbohydrate, Dionex EGC 400 KOH, Dionex CarboPac PA200 1 mm column, Dionex EGC 400 MSA, TN 20, TN 70671, TN20, TN70671 |
| Matrix: | DI water |
| Author: | Beibei Huang and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |