Find methods for your needs
Fast methods for the determination of ibuprofen in drug products
Description
This application note demonstrates straightforward approaches to improve the throughput compared with the USP 40 method by using a 50 × 2.1 mm column, with a 2.6 μm particle size, operated in a UHPLC instrument (Thermo Scientific™ Vanquish™ Horizon UHPLC system), while maintaining USP acceptance criteria on relative retention time, resolution, and tailing factors. The UHPLC system was run with Empower 3Market: | Pharma |
Keywords: | Ibuprofen, Pharmaceutical, Valerophenone, Chromeleon CDS, Drug, DII for Empower software, USP 40 monograph, Empower 3 |
Matrix: | Tablet |
Author: | Sylvia Grosse, Mauro De Pra, Frank Steiner |
Affiliation: | Thermo Fisher Scientific, Germering, Germany |
For Research Use Only. Not for use in diagnostic procedures.