Find methods for your needs
Determination of ultratrace elements in semiconductor grade Isopropyl Alcohol using the Thermo Scientific iCAP TQ ICP-MS
Description
To determine ultratrace metal concentrations in semiconductor grade isopropyl alcohol (IPA). Use cold plasma and triple quadrupole technologies to reduce background equivalent concentrations (BEC) and detection limits (LoD). Demonstrate the use of the Thermo Scientific™ iCAP™ TQ ICP-MS to perform reproducible ultratrace ng·L⁻¹ (ppt) measurements with reliable switching between multiple analysis modes (hot/cold plasma, single/ triple quadrupole) within a single measurement.Market: | Electronics & Semiconductors |
Keywords: | IPA, semiconductor, Triple Quadrupole, Cold plasma, iCAP TQ, ultratrace |
Matrix: | IPA |
Author: | Tomoko Vincent |
Affiliation: | Thermo Fisher Scientific, Bremen, Germany |
For Research Use Only. Not for use in diagnostic procedures.