Find methods for your needs
Analysis of plating baths using the Thermo Scientific iCAP 7400 ICP-OES
Description
This application note describes the performance of the Thermo Scientific iCAP 7400 ICP-OES Duo analyzing different elements in different types of plating baths. The duo viewing offers optimal method conditions using axial view for traces and radial view for major elements.| Market: | Material Science; Chemical |
| Keywords: | Method of standard addition, Plating baths, Sulphuric acid levels |
| Matrix: | Chromium, Chromium and aluminium, Chromium and ammonium fluoride, Nickel |
| Author: | Nora Bartsch |
| Affiliation: | Thermo Fisher Scientific, Bremen, Germany |
For Research Use Only. Not for use in diagnostic procedures.