Find methods for your needs
AN72693: Determination of Total Fluorine, Chlorine, and Sulfur in Aromatic Hydrocarbons by Oxidative Pyrolytic Combustion Followed by Ion Chromatography Be the first to rate this application
Description
This application note describes determination of fluorine, chlorine, and sulfur in aromatic hydrocarbon samples based on ASTM method D7359-14a. A gasoline sample is used here as an example of an aromatic hydrocarbon sample to demonstrate method capabilities. The CIC system used for this method includes a Mitsubishi™ Automatic Combustion Unit Model AQF-2100H system and a Thermo Scientific™ Dionex™ Integrion™ HPIC™ compact IC system equipped with the Thermo Scientific™ Dionex™ IonPac™ AS18-4μm column set. Results for linearity, precision, and accuracy are described here.
Market: Environmental
Keywords: Dionex IonPac AS18-4μm Column, HPIC, CIC, Gasoline, ASTM method D7359-14a, Dionex Integrion, suppressed conductivity detection, TN72211, AN 72693, TN000767, AN72693
Matrix: Gasoline
Author: Sachin Patil and Jeffrey Rohrer
Affiliation: Thermo Fisher Scientific Sunnyvale, CA
Uploaded on 5/7/2018.
For Research Use Only. Not for use in diagnostic procedures.