Find methods for your needs
AU72648: Determination of Gentamicin and Related Impurities in Gentamicin Sulfate using Simple Eluents (Part B)
Description
This application update demonstrates that gentamicin sulfate and related impurities can be separated quickly compared to the method in the United States Pharmacopeia (USP) Gentamicin Sulfate monograph using a Dionex IonPac AmG-3µm C18 column with 100 mM TFA (trifluoroacetic acid) with 2% acetonitrile as the eluent . (Method B in AU 72648)). This method meets or exceeds the performance criteria for Content of Gentamicins in the USP Gentamicin Sulfate monograph.| Market: | Pharma |
| Keywords: | aminoglycoside, Antibiotic, Drug Substance, Electrochemical detection, USP, PAD, Dionex ICS 5000+ HPIC system, Dionex ICS-6000, Dionex IonPac AmG-3µm C18 column, EP, ion pair reversed phase HPLC |
| Matrix: | Water |
| Author: | Jingli Hu and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.