Find methods for your needs
AU72648: Determination of Gentamicin and Related Impurities in Gentamicin Sulfate using Simple Eluents (Part A)
Description
This application update demonstrates that gentamicin sulfate and related impurities can be separated with a Dionex IonPac AmG-3µm C18 column using a simple eluent method (100 mM TFA (trifluoroacetic acid) and no organic solvent. (Method A in AU 72648)). This method meets or exceeds the performance criteria for Content of Gentamicins in the United States Pharmacopeia (USP) Gentamicin Sulfate monograph.| Market: | Pharma |
| Keywords: | aminoglycoside, Antibiotic, Drug Substance, Electrochemical detection, USP, PAD, Dionex ICS 5000+ HPIC system, Dionex ICS-6000, Dionex IonPac AmG-3µm C18 column, EP, ion pair reversed phase HPLC |
| Matrix: | Water |
| Author: | Jingli Hu and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.