Find methods for your needs
AN72609: Using IC with Electrospray Ionization Mass Spectrometry for the Determination of Cations and Amines in Alkanolamine Scrubbing Solutions: Synthetic Sample Be the first to rate this application
Description
Alkanolamines, such as methyldiethanolamine (MDEA), diethanolamine (DEA), and ethanolamine (EA), are used to neutralize the sour gas impurities. The amine solutions are monitored for concentrations of alkanolamine and inorganic cations to maintain neutralizing capacity and to minimize salt-induced corrosion, respectively. In this application note, µg/L to mg/L concentrations of sodium, ammonium, and calcium in high concentrations (mg/L to percent) of alkanolamines are determined by IC-MS on the Integrion IC and ISQ EC mass spectrometer. See also TN72611 for configuration.
Market: Petrochem, Oil and Gas; Environmental
Keywords: Diethanolamine, Ethanolamine, heat stable salts, sour gas, RFIC, DEA, IC-MS, triethanolamine, Methyldiethanolamine, Reagent-Free IC, Ammonium, cations, Integrion, ISQ EC mass spectrometer, Monoethanolamine, electrospray, IC-ESI MS, MDEA, IonPac CS19 column, ISQ EC, EA, TN72611
Matrix: 1000-fold diluted diethanolamine (DEA)
Author: Terri Christison and Jeff Rohrer
Affiliation: Thermo Fisher Scientific
Uploaded on 4/10/2018.
For Research Use Only. Not for use in diagnostic procedures.