Find methods for your needs
AN72609: Using Ion Chromatography with Electrospray Ionization Mass Spectrometry for the Determination of Cations and Amines in Alkanolamine Scrubbing Solutions: MDEA
Description
Alkanolamines, such as methyldiethanolamine (MDEA), diethanolamine (DEA), and ethanolamine (EA), are used to neutralize the sour gas impurities. The amine solutions are monitored for concentrations of alkanolamine and inorganic cations to maintain neutralizing capacity and to minimize salt-induced corrosion, respectively. In this application note, µg/L to mg/L concentrations of sodium, ammonium, and calcium in high concentrations (mg/L to percent) of alkanolamines are determined by IC-MS on the Integrion IC and ISQ EC mass spectrometer. See also TN72611 for configuration.| Market: | Petrochem, Oil and Gas; Environmental |
| Keywords: | Diethanolamine, Ethanolamine, heat stable salts, sour gas, RFIC, DEA, IC-MS, triethanolamine, Methyldiethanolamine, Reagent-Free IC, Ammonium, cations, Integrion, ISQ EC mass spectrometer, Monoethanolamine, electrospray, IC-ESI MS, MDEA, IonPac CS19 column, ISQ EC, EA, TN72611 |
| Matrix: | methyldiethanolamine (MDEA) scrubbing amine sol. |
| Author: | Terri Christison and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.