Find methods for your needs
Improved analysis of organic acids using a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of organic acids. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column, with UV detection at 280 nm.| Market: | Chemical |
| Keywords: | 3,4-Dihydroxy phenyl acetic acid, Caffeic acid, Coumaric acid, Ferulic acid, HPLC, Hydroxybenzoic acid, Hypersil GOLD, Organic acids, Salicylic acid, UV detection, Vanillic acid |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.