Find methods for your needs
Determination of trace elements in steels and alloys using the Thermo Scientific iCAP 7400 ICP-OES
Description
The rapid, precise and accurate determination of minor and trace elements in steels and iron alloys is analytically demanding, but of great commercial significance. This note demonstrates that closed vessel microwave digestion, followed by analysis using the Thermo Scientific iCAP 7000 Plus Series ICP-OES, provides a cost effective solution.Market: | Metals & Mining; Chemical |
Keywords: | Microwave digestion, Alloy, Steel |
Matrix: | steels and iron alloys |
Author: | Sanja Asendorf, Application Specialist |
Affiliation: | Thermo Fisher Scientific (Bremen) GmbH |
For Research Use Only. Not for use in diagnostic procedures.