Find methods for your needs
Low level quantification of PCDD/Fs in animal feed using the Thermo Scientific TSQ 9000 triple quadrupole GC-MS/MS system with AEI source
Description
The performance of the TSQ 9000 triple quadrupole GC-MS/MS system equipped with a new advanced electron ionization (AEI) source was evaluated for the analysis of PCDD/Fs in solvent standards and real food/feedstuff samples.| Market: | Food and Beverage |
| Keywords: | sensitivity, GC-MS/MS, PCDD, PCDF, Persistent Organic Pollutants (POPs), advanced electron ionization, AEI, TSQ 9000, confirmatory analysis, Dionex ASE 350 Accelerated Solvent Extractor, polychlorinated dibenzo-p-dioxins, polychlorinated dibenzo-p-furans |
| Matrix: | Alfalfa, Grass, Pork fat, Premix, Sheep meat |
| Author: | Richard Law |
For Research Use Only. Not for use in diagnostic procedures.