Find methods for your needs
AN72632: Fast and Sensitive Determination of Lactose in Lactose-Free Products using HPAE-PAD
Description
The increased market demand for lactose-free products has created a need for a fast, reliable, and sensitive method to analyze these products. HPAE-PAD is one of the most powerful techniques for carbohydrate determinations. It is a well-established technique for lactose and lactulose determinations. In this work, we developed an HPAE-PAD method using the recently introduced Thermo Scientific™ Dionex™ CarboPac™ PA210-4µm column to separate lactose from other structurally similar milk sugars. Here we determined lactose in eleven lactose-free products (both dairy and baked) using HPAE-PAD.| Market: | Food and Beverage |
| Keywords: | Dionex, Dionex ICS-5000+, Electrochemical detection, Glucose, Lactulose, Milk, Lactose, Raffinose, galactose, HPAE-PAD, dairy products, Chocolate, Epilactose, ICS-6000, baked goods, cookies, allolactose, TN 20, TN20, TN70671, AN 72632, TN 70671 CarboPac PA210 4μm column |
| Matrix: | baked products, Dairy sample |
| Author: | Manali Aggrawal, Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |