Find methods for your needs
Ultra low level quantification of pesticides in baby foods using an advanced triple quadrupole GC-MS/MS system
Description
In this study, the quantitative performance of the Thermo Scientific™ TSQ™ 9000 triple quadrupole GC-MS/MS system was assessed for the analysis of more than 200 pesticides in baby food at ultra low concentrations (as low as 0.025 μg/kg).| Market: | Food and Beverage |
| Keywords: | QuEChERS, sensitivity, Baby Food, Food Safety, GC-MS/MS, robustness, advanced electron ionization, AEI, pesticide residue, Programmable Temperature Vaporization, triple quadrupole mass spectrometry, TSQ 9000 |
| Matrix: | Baby food: fruit and vegetable based |
| Author: | Richard Law; Aaron Lamb; Paul; Cristian Cojocariu |
| Affiliation: | Thermo Fisher Scientific, |
For Research Use Only. Not for use in diagnostic procedures.