Find methods for your needs
AN72610: Determination of Carbohydrates in Algal Biofuel Samples (Part B: Long Method)
Description
The current work describes two methods that use a Dionex CarboPac PA20-4µm column for determination key sugars present in acid hydrolyzed algal biomass. The long method separates the two uronic acids in addition to the twelve other sugars separated by the shorter method. When a detailed sugar composition calculation is desired, the long method can be used. The method was validated for precision, accuracy, and robustness during routine use.| Market: | Energy |
| Keywords: | biofuel, Dionex ICS-5000+, Electrochemical detection, HPAE-PAD, Sugar, RFIC System, ICS-6000, Dionex CarboPac PA20-4μm column, microalgae, uronic acid, TN 20, TN 70671, TN20, TN70671, AN 72610 |
| Matrix: | DI water |
| Author: | Sachin Patil and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.