Find methods for your needs
AN72610: Determination of Carbohydrates in Algal Biofuel Samples (Part A: Short Method)
Description
The current work describes two methods that use a Dionex CarboPac PA20-4µm column for determination key sugars present in acid hydrolyzed algal biomass. The short method separates twelve common carbohydrate sugars in less than fifteen minutes. This method does not separate uronic acids, which are key components that are present in low concentrations. The method was validated for precision, accuracy, and robustness during routine use.| Market: | Energy |
| Keywords: | biofuel, Dionex ICS-5000+, Electrochemical detection, HPAE-PAD, Sugar, RFIC System, ICS-6000, Dionex CarboPac PA20-4μm column, microalgae, uronic acid, TN 20, TN 70671, TN20, TN70671, AN 72610 |
| Matrix: | DI water |
| Author: | Sachin Patil and Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.