Find methods for your needs
Analysis of Hydride Forming Elements with the Thermo Scientific iCAP 7000 Plus Series ICP-OES and the Basic Hydride Generator Kit
Description
This application note demonstrates the ability of the Thermo Scientific iCAP 7000 Plus Series ICP-OES to determine hydride forming elements with the Basic Hydride Generator Kit, which significantly improves detection capability, precision and accuracy for arsenic, bismuth, antimony, selenium and mercury compared to standard ICP-OES sample introduction.| Market: | Environmental; Water Analysis; Food and Beverage |
| Keywords: | Accuracy, Hydride, Precision, Trace Elemental |
| Matrix: | 0.5% sodium borohydride, 0.5% sodium hydroxide, 1% potassium iodide, 10% hydrochloric acid |
| Author: | Nora Bartsch |
| Affiliation: | Thermo Fisher Scientific, Bremen, Germany |
For Research Use Only. Not for use in diagnostic procedures.