Find methods for your needs
Quantification of paclitaxel, its degradants and related substances using UHPLC with charged aerosol detection
Description
This application shows the quantification of multiple impurities with a single calibrant by taking advantage of the uniform response of the charged aerosol detector (CAD). The inverse gradient compensation wizard is also highlighted.| Market: | Pharma |
| Keywords: | Paclitaxel, Taxane, Charged aerosol detection, Impurity analysis, Vanquish Flex Duo UHPLC, forced degradation, global calibration, inverse gradient, stability analysis, Taxol |
| Matrix: | Methanol |
| Author: | Menz, Michael(1); Steiner, Frank(1); Acworth, Ian(2) |
| Affiliation: | (1) Thermo Fisher Scientific, Germering, Germany; (2) Thermo Fisher Scientific, Chelmsford, MA, USA |