Find methods for your needs
Simultaneous quantification of nevirapine and low-level impurities Be the first to rate this application
Description
This application describes the straightforward simultaneous quantification of the active pharmaceutical ingredient Nevirapine and its low-level impurities from the same run. The separation takes just 2.8 min und the highly linear and low-noise performance of the Thermo Scientific Vanquish DAD FG ensures API and impurity quantification under compliance with ICH guidelines within one injection.
Market: Pharma
Keywords: API, Impurity, Active pharmaceutical ingredient, linear range, ICH guidelines
Matrix: mobile Phase
Author: Maria Grübner, Frank Steiner, Holger Franz
Affiliation: Thermo Fisher Scientific, Germering, Germany
Uploaded on 2/21/2018.
For Research Use Only. Not for use in diagnostic procedures.